siRNA Details
Gene Symbol: KIAA0100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014680.3 | NP_055495.2 | 6 | 653 | ||
XM_005258073.2 | XP_005258130.1 | 6 | 648 | |||
XM_017025423.1 | XP_016880912.1 | 5 | 502 | |||
GenBank mRNA | AB621823.1 | - | 6 | 650 | ||
AK130007.1 | - | 6 | 608 | |||
AK307216.1 | - | 5 | 592 | |||
AY943906.1 | - | 3 | 498 | |||
BC048096.1 | - | 5 | 903 | |||
BC050440.1 | - | 6 | 649 | |||
BC120873.1 | - | 6 | 512 | 40075102 | ||
CR933634.1 | - | 6 | 605 | |||
D43947.3 | - | 6 | 625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
