siRNA Details
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030647.1 | NP_085150.1 | 10 | 1281 | ||
XM_011516587.2 | XP_011514889.1 | 4 | 306 | |||
GenBank mRNA | AK131497.1 | - | 10 | 1282 | ||
BX641017.1 | - | 2 | 61 | |||
BX649115.1 | - | 5 | 548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
