siRNA Details
Gene Symbol: SLC45A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080431.2 | NP_001073900.1 | 6 | 1909 | ||
NM_001286646.1 | NP_001273575.1 | 6 | 2154 | |||
NM_001286648.1 | NP_001273577.1 | 6 | 1909 | |||
XM_011517169.2 | XP_011515471.1 | 7 | 2195 | |||
XM_011517170.2 | XP_011515472.1 | 8 | 3000 | |||
XM_011517171.2 | XP_011515473.1 | 7 | 2250 | |||
XM_011517172.2 | XP_011515474.1 | 7 | 2739 | |||
XM_011517173.2 | XP_011515475.1 | 7 | 2195 | |||
XM_011517174.2 | XP_011515476.1 | 6 | 1909 | |||
XM_011517176.2 | XP_011515478.1 | 6 | 2006 | |||
XM_011517177.2 | XP_011515479.1 | 6 | 2447 | |||
XM_011517178.2 | XP_011515480.1 | 6 | 4624 | |||
XM_011517179.2 | XP_011515481.1 | 6 | 1739 | |||
XM_017013648.1 | XP_016869137.1 | 7 | 3274 | |||
XM_017013649.1 | XP_016869138.1 | 7 | 2179 | |||
XM_017013650.1 | XP_016869139.1 | 6 | 2641 | |||
GenBank mRNA | AB032952.1 | - | 4 | 1305 | ||
AK128212.1 | - | 6 | 1909 | |||
BC090891.1 | - | 6 | 1670 | |||
BC157840.1 | - | 6 | 1651 | 9057059 | ||
BC171836.1 | - | 6 | 1651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
