siRNA Details
Gene Symbol: SLC45A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080431.2 | NP_001073900.1 | 8 | 3205 | ||
NM_001286646.1 | NP_001273575.1 | 8 | 3457 | |||
XM_011517169.2 | XP_011515471.1 | 9 | 3498 | |||
XM_011517170.2 | XP_011515472.1 | 10 | 4303 | |||
XM_011517171.2 | XP_011515473.1 | 9 | 3553 | |||
XM_011517172.2 | XP_011515474.1 | 9 | 4042 | |||
XM_011517174.2 | XP_011515476.1 | 8 | 3212 | |||
XM_011517176.2 | XP_011515478.1 | 8 | 3309 | |||
XM_011517177.2 | XP_011515479.1 | 8 | 3750 | |||
XM_011517178.2 | XP_011515480.1 | 8 | 5927 | |||
XM_011517179.2 | XP_011515481.1 | 8 | 3042 | |||
XM_017013648.1 | XP_016869137.1 | 9 | 4577 | |||
XM_017013649.1 | XP_016869138.1 | 9 | 3482 | |||
XM_017013650.1 | XP_016869139.1 | 8 | 3944 | |||
GenBank mRNA | AB032952.1 | - | 5 | 3108 | ||
AK128212.1 | - | 8 | 3205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
