siRNA Details
Gene Symbol: SLC45A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080431.2 | NP_001073900.1 | 8 | 4790 | ||
NM_001286646.1 | NP_001273575.1 | 8 | 5042 | |||
XM_011517169.2 | XP_011515471.1 | 9 | 5083 | |||
XM_011517170.2 | XP_011515472.1 | 10 | 5888 | |||
XM_011517171.2 | XP_011515473.1 | 9 | 5138 | |||
XM_011517172.2 | XP_011515474.1 | 9 | 5627 | |||
XM_011517174.2 | XP_011515476.1 | 8 | 4797 | |||
XM_011517176.2 | XP_011515478.1 | 8 | 4894 | |||
XM_011517177.2 | XP_011515479.1 | 8 | 5335 | |||
XM_011517178.2 | XP_011515480.1 | 8 | 7512 | |||
XM_011517179.2 | XP_011515481.1 | 8 | 4627 | |||
XM_017013648.1 | XP_016869137.1 | 9 | 6162 | |||
XM_017013649.1 | XP_016869138.1 | 9 | 5067 | |||
XM_017013650.1 | XP_016869139.1 | 8 | 5529 | |||
GenBank mRNA | AB032952.1 | - | 5 | 4691 | ||
AL137490.1 | - | 1 | 738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
