siRNA Details
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148894.2 | NP_683692.2 | 16 | 8422 | ||
XM_005248150.3 | XP_005248207.1 | 16 | 8439 | |||
XM_005248151.3 | XP_005248208.1 | 16 | 8439 | |||
XM_006713958.3 | XP_006714021.1 | 16 | 8439 | |||
XM_011513827.2 | XP_011512129.1 | 16 | 8439 | |||
XM_011513829.2 | XP_011512131.1 | 16 | 8439 | |||
XM_011513830.2 | XP_011512132.1 | 14 | 7822 | |||
XM_017008009.1 | XP_016863498.1 | 16 | 8439 | |||
XM_017008010.1 | XP_016863499.1 | 16 | 8439 | |||
GenBank mRNA | AB037748.2 | - | 7 | 4566 | ||
AK025361.1 | - | 2 | 83 | |||
AK122806.1 | - | 4 | 1755 | |||
AK298130.1 | - | 7 | 1797 | |||
AL137350.1 | - | 7 | 977 | |||
BC016987.1 | - | 6 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
