siRNA Details
Gene Symbol: PPP1R3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184745.1 | NP_001171674.1 | 4 | 473 | ||
NM_033215.4 | NP_149992.3 | 4 | 1416 | |||
XM_005272687.3 | XP_005272744.1 | 4 | 1434 | |||
XM_017029932.1 | XP_016885421.1 | 4 | 5844 | |||
XM_017029933.1 | XP_016885422.1 | 4 | 5841 | |||
GenBank mRNA | AK056297.1 | - | 3 | 833 | ||
AK056909.1 | - | 4 | 1241 | |||
AK127066.1 | - | 1 | 2074 | |||
AK303738.1 | - | 5 | 739 | |||
BC131588.1 | - | 4 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
