siRNA Details
Gene Symbol: KIAA2022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008537.2 | NP_001008537.1 | 3 | 2847 | ||
GenBank mRNA | AB095942.1 | - | 4 | 2475 | ||
AK128651.1 | - | 1 | 1839 | |||
AK307471.1 | - | 3 | 2841 | |||
AY563507.1 | - | 3 | 2544 | |||
BC152557.1 | - | 4 | 2475 | 8860127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
