siRNA Details
Gene Symbol: DNAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303270.1 | NP_001290199.1 | 12 | 1758 | ||
NM_020877.3 | NP_065928.2 | 10 | 2886 | |||
XM_011523663.1 | XP_011521965.1 | 12 | 1754 | |||
XM_011523664.2 | XP_011521966.1 | 13 | 1903 | |||
XM_011523666.1 | XP_011521968.1 | 12 | 1754 | |||
XM_011523667.2 | XP_011521969.1 | 11 | 1657 | |||
XM_011523668.1 | XP_011521970.1 | 2 | 137 | |||
XM_011523670.2 | XP_011521972.1 | 12 | 1754 | |||
XM_017024219.1 | XP_016879708.1 | 12 | 1754 | |||
GenBank mRNA | AB040936.2 | - | 10 | 2886 | ||
AK292607.1 | - | 12 | 1754 | |||
BC034225.1 | - | 12 | 1757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
