siRNA Details
Gene Symbol: ZNF275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080485.3 | NP_001073954.3 | 5 | 1527 | ||
GenBank mRNA | AK122976.1 | - | 5 | 1527 | ||
AK127133.1 | - | 1 | 1323 | |||
BC001545.1 | - | 3 | 2270 | |||
BC136634.1 | - | 2 | 2651 | 9020624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
