siRNA Details
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 2 | 331 | ||
GenBank mRNA | AB449912.1 | - | 2 | 220 | ||
AK293521.1 | - | 2 | 289 | |||
AK293875.1 | - | 2 | 290 | |||
AK294776.1 | - | 2 | 158 | |||
AK298884.1 | - | 2 | 318 | |||
BC052609.1 | - | 2 | 276 | |||
BC082255.1 | - | 2 | 253 | |||
BC101817.1 | - | 2 | 257 | 8069323 | ||
BC101819.1 | - | 2 | 257 | 8069325 | ||
BC125088.1 | - | 2 | 246 | 40117866 | ||
BC125089.1 | - | 2 | 246 | |||
DA087200.1 | - | 2 | 331 | |||
L08240.1 | - | 1 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
