siRNA Details
Gene Symbol: ANO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020959.2 | NP_066010.1 | 10, 11 | 1432 | ||
XM_017027048.1 | XP_016882537.1 | 10, 11 | 1437 | |||
GenBank mRNA | AB046843.2 | - | 10, 11 | 1432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
