siRNA Details
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080429.2 | NP_001073898.1 | 13 | 2048 | ||
NM_020902.1 | NP_065953.1 | 11 | 1967 | |||
XM_005272491.3 | XP_005272548.1 | 12 | 1984 | |||
XM_011528154.2 | XP_011526456.1 | 13 | 1967 | |||
XM_017027036.1 | XP_016882525.1 | 13 | 2021 | |||
GenBank mRNA | AB040976.1 | - | 4 | 870 | ||
AL833927.1 | - | 1 | 650 | |||
BC020431.1 | - | 13 | 1891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
