siRNA Details
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080429.2 | NP_001073898.1 | 19 | 3792 | ||
NM_020902.1 | NP_065953.1 | 17 | 3711 | |||
XM_005272491.3 | XP_005272548.1 | 18 | 3728 | |||
XM_011528154.2 | XP_011526456.1 | 19 | 3711 | |||
XM_017027036.1 | XP_016882525.1 | 19 | 3778 | |||
GenBank mRNA | AB040976.1 | - | 10 | 2614 | ||
AF289580.1 | - | 7 | 1664 | |||
AL833927.1 | - | 7 | 2394 | |||
BC020431.1 | - | 19 | 3635 | |||
BC128201.1 | - | 3 | 310 | |||
BC128202.1 | - | 3 | 310 |

More Information
Other Species Targeted

Gene Ontology Categories:
