siRNA Details
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080429.2 | NP_001073898.1 | 15 | 3162 | ||
NM_020902.1 | NP_065953.1 | 13 | 3081 | |||
XM_005272491.3 | XP_005272548.1 | 14 | 3098 | |||
XM_011528154.2 | XP_011526456.1 | 15 | 3081 | |||
XM_017027036.1 | XP_016882525.1 | 15 | 3135 | |||
GenBank mRNA | AB040976.1 | - | 6 | 1984 | ||
AF289580.1 | - | 3 | 1032 | |||
AL833927.1 | - | 3 | 1764 | |||
BC020431.1 | - | 15 | 3005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
