siRNA Details
Gene Symbol: KIAA1107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000800.1 | XP_016856289.1 | 4 | 464 | ||
GenBank mRNA | AB029030.1 | - | 3 | 235 | ||
AK302496.1 | - | 3 | 340 | |||
DC331933.1 | - | 3 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
