siRNA Details
Gene Symbol: MTMR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004686.4 | NP_004677.3 | 9, 10 | 1338 | ||
XM_006716414.3 | XP_006716477.2 | 4, 5 | 1704 | |||
XM_011544692.2 | XP_011542994.1 | 6, 7 | 777 | |||
XM_011544693.2 | XP_011542995.1 | 5, 6 | 1239 | |||
XM_017013956.1 | XP_016869445.1 | 8, 9 | 1027 | |||
GenBank mRNA | AF073482.1 | - | 5, 6 | 607 | ||
AK294468.1 | - | 9, 10 | 1099 | |||
AK303762.1 | - | 1, 2 | 161 | |||
AK316022.1 | - | 9, 10 | 1148 | |||
AK316283.1 | - | 9, 10 | 1158 | |||
BC130577.1 | - | 9, 10 | 1124 | 40146608 | ||
BC130579.1 | - | 9, 10 | 1124 | 40146610 | ||
CR749240.1 | - | 9, 10 | 1136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
