siRNA Details
Gene Symbol: MTMR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004686.4 | NP_004677.3 | 11 | 1459 | ||
XM_006716414.3 | XP_006716477.2 | 6 | 1825 | |||
XM_011544692.2 | XP_011542994.1 | 8 | 898 | |||
XM_011544693.2 | XP_011542995.1 | 7 | 1360 | |||
XM_017013956.1 | XP_016869445.1 | 10 | 1148 | |||
GenBank mRNA | AF073482.1 | - | 7 | 728 | ||
AK294468.1 | - | 11 | 1220 | |||
AK303762.1 | - | 3 | 282 | |||
AK303803.1 | - | 2 | 232 | |||
AK316022.1 | - | 11 | 1269 | |||
AK316283.1 | - | 11 | 1279 | |||
BC130577.1 | - | 11 | 1245 | 40146608 | ||
BC130579.1 | - | 11 | 1245 | 40146610 | ||
CR749240.1 | - | 11 | 1257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
