siRNA Details
Gene Symbol: CRYBG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153605.3 | NP_705833.3 | 16 | 8201 | ||
XM_005247117.4 | XP_005247174.1 | 15 | 7904 | |||
GenBank mRNA | AF451988.1 | - | 4 | 330 | ||
AK091199.1 | - | 8 | 672 | |||
AL831927.1 | - | 4 | 224 | |||
BX647079.1 | - | 8 | 689 | |||
BX648353.1 | - | 13 | 2251 | |||
CR749313.1 | - | 13 | 1326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
