siRNA Details
Gene Symbol: CRYBG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153605.3 | NP_705833.3 | 18, 19 | 8548 | ||
XM_005247117.4 | XP_005247174.1 | 17, 18 | 8251 | |||
GenBank mRNA | AF451988.1 | - | 6, 7 | 665 | ||
AK091199.1 | - | 10, 11 | 1019 | |||
AK098569.1 | - | 2, 3 | 340 | |||
AL831927.1 | - | 6, 7 | 571 | |||
BC036247.1 | - | 1, 2 | 91 | |||
BC050394.1 | - | 3, 4 | 331 | |||
BX647079.1 | - | 10, 11 | 1036 | |||
BX648353.1 | - | 15, 16 | 2598 | |||
CR749313.1 | - | 15, 16 | 1673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
