siRNA Details
Gene Symbol: CRYBG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153605.3 | NP_705833.3 | 21 | 8897 | ||
XM_005247117.4 | XP_005247174.1 | 20 | 8600 | |||
GenBank mRNA | AF451988.1 | - | 9 | 1014 | ||
AK091199.1 | - | 13 | 1368 | |||
AK098569.1 | - | 5 | 689 | |||
AL831927.1 | - | 9 | 920 | |||
BC036247.1 | - | 4 | 440 | |||
BC050394.1 | - | 6 | 680 | |||
BX647079.1 | - | 13 | 1385 | |||
BX648353.1 | - | 18 | 2947 | |||
CR749313.1 | - | 18 | 2022 |
Gene Symbol: MINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM979079.1 | - | 1 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
