siRNA Details
Gene Symbol: TMEM131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015348.1 | NP_056163.1 | 12 | 1343 | ||
XM_005263909.1 | XP_005263966.1 | 13 | 1429 | |||
XM_005263910.1 | XP_005263967.1 | 13 | 1429 | |||
XM_005263911.1 | XP_005263968.1 | 12 | 1312 | |||
XM_005263912.3 | XP_005263969.1 | 11 | 1517 | |||
XM_011510869.1 | XP_011509171.1 | 13 | 1429 | |||
GenBank mRNA | D87446.1 | - | 11 | 881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
