siRNA Details
Gene Symbol: TMEM131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015348.1 | NP_056163.1 | 14 | 1598 | ||
XM_005263909.1 | XP_005263966.1 | 15 | 1684 | |||
XM_005263910.1 | XP_005263967.1 | 15 | 1684 | |||
XM_005263911.1 | XP_005263968.1 | 14 | 1567 | |||
XM_005263912.3 | XP_005263969.1 | 13 | 1772 | |||
XM_011510869.1 | XP_011509171.1 | 15 | 1684 | |||
GenBank mRNA | D87446.1 | - | 13 | 1136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
