siRNA Details
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020853.1 | NP_065904.1 | 8 | 1236 | ||
XM_017019706.1 | XP_016875195.1 | 10 | 1547 | |||
GenBank mRNA | AB040900.1 | - | 5 | 643 | ||
AK125278.1 | - | 8 | 1236 | |||
AK294732.1 | - | 2 | 291 | |||
AL832902.1 | - | 5 | 541 | |||
BC039267.1 | - | 8 | 1225 | 5274878 | ||
BC132772.1 | - | 8 | 1333 | 40146794 | ||
BC132774.1 | - | 8 | 1333 | 40146796 | ||
CR936647.1 | - | 8 | 1227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
