siRNA Details
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020853.1 | NP_065904.1 | 13 | 4605 | ||
GenBank mRNA | AB040900.1 | - | 10 | 4010 | ||
AK125278.1 | - | 14 | 2517 | |||
BC030126.2 | - | 1 | 731 | |||
BC039267.1 | - | 4594 | 5274878 | |||
CR936647.1 | - | 13 | 4594 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
