siRNA Details
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 3 | 721 | ||
XM_005260282.4 | XP_005260339.1 | 3 | 1007 | |||
XM_005260283.4 | XP_005260340.1 | 4 | 1031 | |||
XM_005260284.4 | XP_005260341.1 | 3 | 306 | |||
XM_005260285.4 | XP_005260342.2 | 3 | 1011 | |||
XM_005260286.4 | XP_005260343.1 | 3 | 931 | |||
XM_006723702.3 | XP_006723765.1 | 4 | 398 | |||
XM_006723703.3 | XP_006723766.1 | 3 | 310 | |||
XM_011528562.2 | XP_011526864.1 | 3 | 715 | |||
XM_011528563.1 | XP_011526865.1 | 3 | 715 | |||
XM_017027669.1 | XP_016883158.1 | 3 | 301 | |||
GenBank mRNA | AK097804.1 | - | 5 | 1384 | ||
BQ067609.1 | - | 3 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
