siRNA Details
Gene Symbol: HAUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015302.1 | NP_056117.1 | 19 | 2627 | ||
XM_011526684.2 | XP_011524986.1 | 18 | 2514 | |||
GenBank mRNA | AB020648.1 | - | 19 | 2600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
