siRNA Details
Gene Symbol: ERMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024896.2 | NP_079172.2 | 8 | 1502 | ||
XM_005251587.1 | XP_005251644.1 | 8 | 1473 | |||
XM_017015139.1 | XP_016870628.1 | 9 | 1295 | |||
GenBank mRNA | AB058718.2 | - | 8 | 1379 | ||
AK093217.1 | - | 7 | 869 | |||
AK127218.1 | - | 8 | 1491 | |||
AK172831.1 | - | 2 | 206 | |||
AL834191.1 | - | 7 | 1134 | |||
BC136771.1 | - | 8 | 1476 | 9020761 | ||
BC136773.1 | - | 8 | 1476 | 9020763 | ||
BC144404.1 | - | 8 | 1476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
