siRNA Details
Gene Symbol: ERMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024896.2 | NP_079172.2 | 13 | 2408 | ||
XM_005251587.1 | XP_005251644.1 | 11 | 2013 | |||
XM_017015139.1 | XP_016870628.1 | 14 | 2201 | |||
GenBank mRNA | AB058718.2 | - | 13 | 2285 | ||
AK026962.1 | - | 5 | 714 | |||
AK093217.1 | - | 11 | 1584 | |||
AK127218.1 | - | 13 | 2397 | |||
AK172831.1 | - | 7 | 1112 | |||
AL834191.1 | - | 12 | 2040 | |||
BC031630.1 | - | 5 | 772 | |||
BC136771.1 | - | 13 | 2382 | 9020761 | ||
BC136773.1 | - | 13 | 2382 | 9020763 | ||
BC144404.1 | - | 13 | 2382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
