siRNA Details
Gene Symbol: NCOR1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003678.1 | 1 | 141 | |||
GenBank mRNA | BC132806.1 | - | 1 | 128 | ||
BC132808.1 | - | 1 | 128 |
Gene Symbol: LOC105379511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135512.1 | 2 | 235 |
Gene Symbol: NCOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF087856.1 | - | 1 | 170 | ||
AF303586.1 | - | 1 | 170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
