siRNA Details
Gene Symbol: RNF215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017981.1 | NP_001017981.1 | 5 | 658 | ||
GenBank mRNA | AK293904.1 | - | 4 | 500 | ||
BM476400.1 | - | 5 | 470 | |||
BM806918.1 | - | 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
