siRNA Details
Gene Symbol: SRRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013694.2 | NP_001013716.2 | 3 | 418 | ||
XM_011530178.2 | XP_011528480.1 | 3 | 481 | |||
XM_017028799.1 | XP_016884288.1 | 3 | 430 | |||
GenBank mRNA | AK293916.1 | - | 4 | 398 | ||
BC066962.1 | - | 4 | 401 | 4820477 | ||
DA325985.1 | - | 3 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
