siRNA Details
Gene Symbol: SRRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013694.2 | NP_001013716.2 | 4, 5 | 627 | ||
XM_011530178.2 | XP_011528480.1 | 4, 5 | 690 | |||
GenBank mRNA | AK293916.1 | - | 5, 6 | 607 | ||
BC066962.1 | - | 5, 6 | 610 | 4820477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
