siRNA Details
Gene Symbol: CHADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138481.1 | NP_612490.1 | 3 | 1561 | ||
XM_005261370.3 | XP_005261427.1 | 3 | 1607 | |||
XM_011529932.2 | XP_011528234.1 | 2 | 2225 | |||
XM_011529933.2 | XP_011528235.1 | 2 | 2223 | |||
XM_011529934.2 | XP_011528236.1 | 2 | 2225 | |||
GenBank mRNA | BC040188.1 | - | 3 | 1529 | ||
BC048421.1 | - | 1 | 584 | |||
BC068590.1 | - | 3 | 1561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
