siRNA Details
Gene Symbol: SAMD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010860.1 | NP_001010860.1 | 1 | 1337 | ||
GenBank mRNA | AK093282.1 | - | 1 | 1353 | ||
AK292571.1 | - | 1 | 1623 | |||
BC104834.1 | - | 1 | 1357 | 8143837 | ||
BC104836.1 | - | 1 | 1357 | 8143839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
