siRNA Details
Gene Symbol: PABPC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124756.2 | NP_001118228.1 | 2 | 427 | ||
NR_134983.1 | 2 | 427 | ||||
GenBank mRNA | DC418955.1 | - | 2 | 430 | ||
DR001377.1 | - | 2 | 415 | |||
EU190483.1 | - | 2 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
