siRNA Details
Gene Symbol: SPTLC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018327.2 | NP_060797.2 | 4 | 785 | ||
XM_011529279.1 | XP_011527581.1 | 4 | 553 | |||
XM_017027922.1 | XP_016883411.1 | 4 | 553 | |||
GenBank mRNA | AK300184.1 | - | 4 | 591 | ||
BC150644.1 | - | 4 | 744 | 9057014 | ||
CF994231.1 | - | 4 | 559 | |||
CX870585.1 | - | 3 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
