siRNA Details
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029871.3 | NP_001025042.2 | 3 | 434 | ||
NM_001040007.2 | NP_001035096.1 | 3 | 434 | |||
XM_017027839.1 | XP_016883328.1 | 3 | 362 | |||
GenBank mRNA | AK122609.1 | - | 3 | 431 | ||
DQ355152.1 | - | 3 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
