siRNA Details
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029871.3 | NP_001025042.2 | 5 | 2273 | ||
NM_001040007.2 | NP_001035096.1 | 4 | 2087 | |||
GenBank mRNA | AK122609.1 | - | 5 | 2269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
