siRNA Details
Gene Symbol: XKR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011718.1 | NP_001011718.1 | 3 | 1421 | ||
GenBank mRNA | AY534245.1 | - | 3 | 1421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
