siRNA Details
Gene Symbol: TLDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304783.1 | NP_001291712.1 | 6 | 1895 | ||
NM_080628.2 | NP_542195.1 | 7 | 1991 | |||
GenBank mRNA | AK123544.1 | - | 7 | 1945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
