siRNA Details
Gene Symbol: SPINT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006652.1 | NP_006643.1 | 2 | 126 | ||
GenBank mRNA | AY372172.2 | - | 2 | 126 | ||
BX088813.1 | - | 2 | 76 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
