siRNA Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014643.3 | NP_055458.1 | 7 | 4957 | ||
XM_011526269.1 | XP_011524571.1 | 6 | 4895 | |||
XM_011526270.2 | XP_011524572.1 | 6 | 5158 | |||
XM_011526271.2 | XP_011524573.1 | 7 | 5272 | |||
XM_011526272.2 | XP_011524574.1 | 7 | 4993 | |||
XM_011526273.2 | XP_011524575.1 | 6 | 4908 | |||
XM_011526274.2 | XP_011524576.1 | 6 | 4914 | |||
XM_011526275.2 | XP_011524577.1 | 6 | 5771 | |||
XM_011526276.1 | XP_011524578.1 | 5 | 4827 | |||
XM_017026097.1 | XP_016881586.1 | 7 | 5028 | |||
GenBank mRNA | D86975.1 | - | 7 | 4957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
