siRNA Details
Gene Symbol: ZC3H12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010888.3 | NP_001010888.3 | 5 | 2171 | ||
XM_011530939.2 | XP_011529241.1 | 6 | 2416 | |||
XM_017029478.1 | XP_016884967.1 | 11 | 3682 | |||
XM_017029479.1 | XP_016884968.1 | 13 | 6975 | |||
XM_017029480.1 | XP_016884969.1 | 11 | 3716 | |||
XM_017029481.1 | XP_016884970.1 | 11 | 3711 | |||
XM_017029482.1 | XP_016884971.1 | 10 | 3621 | |||
XM_017029483.1 | XP_016884972.1 | 9 | 3417 | |||
XM_017029484.1 | XP_016884973.1 | 9 | 3049 | |||
GenBank mRNA | BC140767.1 | - | 5 | 2171 | 9021635 | |
BC171781.1 | - | 5 | 2171 | 9054435 | ||
BX647241.1 | - | 7 | 1921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
