siRNA Details
Gene Symbol: UBE2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012989.2 | NP_001013007.1 | 1 | 199 | ||
NR_121210.1 | 1 | 199 | ||||
GenBank mRNA | GU727624.1 | - | 1 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
