siRNA Details
Gene Symbol: MAP7D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173516.1 | NP_001166987.1 | 11 | 1920 | ||
NM_001173517.1 | NP_001166988.1 | 10 | 1927 | |||
NM_024597.3 | NP_078873.2 | 11 | 2032 | |||
XM_005262472.1 | XP_005262529.1 | 11 | 1884 | |||
XM_017029843.1 | XP_016885332.1 | 11 | 1884 | |||
GenBank mRNA | AK301478.1 | - | 11 | 1921 | ||
AL832120.1 | - | 10 | 1719 | |||
AL832475.1 | - | 1 | 35 | |||
BC040518.1 | - | 11 | 1896 | |||
BC064350.1 | - | 11 | 1712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
