siRNA Details
Gene Symbol: HUWE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031407.6 | NP_113584.3 | 9 | 997 | ||
XM_005261965.3 | XP_005262022.1 | 8 | 1064 | |||
XM_011530747.1 | XP_011529049.1 | 9 | 991 | |||
XM_011530751.2 | XP_011529053.1 | 8 | 859 | |||
XM_017029191.1 | XP_016884680.1 | 7 | 3722 | |||
XM_017029192.1 | XP_016884681.1 | 7 | 3722 | |||
XM_017029193.1 | XP_016884682.1 | 7 | 3722 | |||
XM_017029194.1 | XP_016884683.1 | 7 | 3722 | |||
XM_017029195.1 | XP_016884684.1 | 7 | 3722 | |||
XM_017029196.1 | XP_016884685.1 | 7 | 3722 | |||
XM_017029197.1 | XP_016884686.1 | 7 | 3722 | |||
XM_017029198.1 | XP_016884687.1 | 8 | 1064 | |||
XM_017029199.1 | XP_016884688.1 | 9 | 991 | |||
XM_017029200.1 | XP_016884689.1 | 8 | 861 | |||
XM_017029201.1 | XP_016884690.1 | 8 | 878 | |||
XM_017029202.1 | XP_016884691.1 | 8 | 854 | |||
XM_017029203.1 | XP_016884692.1 | 8 | 860 | |||
XM_017029204.1 | XP_016884693.1 | 7 | 3721 | |||
XM_017029205.1 | XP_016884694.1 | 7 | 3722 | |||
XM_017029206.1 | XP_016884695.1 | 7 | 3722 | |||
GenBank mRNA | AY772009.1 | - | 7 | 642 | ||
AY929612.1 | - | 6 | 594 | |||
BC018750.1 | - | 9 | 996 | |||
BC038184.1 | - | 9 | 996 | |||
DQ097177.1 | - | 9 | 996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
