siRNA Details
Gene Symbol: NDFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161407.1 | NP_001154879.1 | 8 | 1358 | ||
NM_019080.2 | NP_061953.2 | 8 | 1418 | |||
XM_006719840.3 | XP_006719903.1 | 8 | 1817 | |||
GenBank mRNA | AB032991.1 | - | 8 | 1182 | ||
AB097019.1 | - | 8 | 1390 | |||
AB097028.1 | - | 8 | 1390 | |||
AB097029.1 | - | 8 | 1390 | |||
AB097030.1 | - | 8 | 1068 | |||
AB097031.1 | - | 8 | 1068 | |||
AK001723.1 | - | 8 | 1209 | |||
AK098708.1 | - | 7 | 979 | |||
AK294839.1 | - | 8 | 1029 | |||
BC021988.1 | - | 8 | 1150 | 4101140 | ||
BC026126.2 | - | 8 | 1179 | 3027907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
