siRNA Details
Gene Symbol: NDFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161407.1 | NP_001154879.1 | 2 | 503 | ||
NM_019080.2 | NP_061953.2 | 2 | 503 | |||
XM_006719840.3 | XP_006719903.1 | 2 | 902 | |||
GenBank mRNA | AB032991.1 | - | 2 | 267 | ||
AB097019.1 | - | 2 | 475 | |||
AB097028.1 | - | 2 | 475 | |||
AB097029.1 | - | 2 | 475 | |||
AB097030.1 | - | 2 | 153 | |||
AB097031.1 | - | 2 | 153 | |||
AK001723.1 | - | 2 | 294 | |||
AK098708.1 | - | 3 | 256 | |||
AK294839.1 | - | 2 | 174 | |||
BC021988.1 | - | 2 | 235 | 4101140 | ||
BC026126.2 | - | 2 | 264 | 3027907 | ||
DC320871.1 | - | 2 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
