siRNA Details
Gene Symbol: SNX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032167.4 | NP_115543.3 | 20 | 2285 | ||
XM_005255682.3 | XP_005255739.1 | 20 | 2310 | |||
XM_011522738.2 | XP_011521040.1 | 21 | 2568 | |||
XM_011522741.2 | XP_011521043.1 | 21 | 2552 | |||
XM_011522743.2 | XP_011521045.1 | 21 | 2556 | |||
XM_011522744.2 | XP_011521046.1 | 20 | 2316 | |||
XM_017023871.1 | XP_016879360.1 | 21 | 2538 | |||
XM_017023872.1 | XP_016879361.1 | 21 | 2385 | |||
XM_017023873.1 | XP_016879362.1 | 20 | 2441 | |||
XM_017023874.1 | XP_016879363.1 | 20 | 2274 | |||
XM_017023875.1 | XP_016879364.1 | 19 | 2092 | |||
XM_017023878.1 | XP_016879367.1 | 11 | 1890 | |||
GenBank mRNA | AK074072.1 | - | 10 | 892 | ||
BC029857.1 | - | 8 | 679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
